Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MADD SIRNA Assays ›
Gene Symbol:
MADD
Gene Name:
MAP kinase activating death domain
Gene Aliases:
DENN, IG20, RAB3GEP
Chromosome Location:
Chr.11: 47269376 - 47330031 on Build GRCh38
UniGene ID:
Hs.82548
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139308
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MADD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135943.1
NP_001129415.1
4
888
NM_001135944.1
NP_001129416.1
4
864
NM_003682.3
NP_003673.3
4
864
NM_130470.2
NP_569826.2
4
1030
NM_130471.2
NP_569827.2
4
864
NM_130472.2
NP_569828.2
4
864
NM_130473.2
NP_569829.2
4
864
NM_130474.2
NP_569830.2
4
864
NM_130475.2
NP_569831.1
4
888
NM_130476.2
NP_569832.2
4
888
XM_005253189.2
XP_005253246.1
4
888
XM_005253196.2
XP_005253253.1
4
888
XM_005253199.2
XP_005253256.1
4
888
XM_005253200.2
XP_005253257.1
4
888
XM_005253201.2
XP_005253258.1
4
888
XM_005253203.2
XP_005253260.1
4
888
XM_005253204.2
XP_005253261.1
4
888
XM_005253205.1
XP_005253262.1
4
888
XM_017018478.1
XP_016873967.1
4
888
XM_017018479.1
XP_016873968.1
4
888
XM_017018480.1
XP_016873969.1
4
888
XM_017018481.1
XP_016873970.1
4
888
XM_017018482.1
XP_016873971.1
4
888
XM_017018483.1
XP_016873972.1
4
888
XM_017018484.1
XP_016873973.1
4
888
XM_017018485.1
XP_016873974.1
4
849
XM_017018486.1
XP_016873975.1
4
888
XM_017018487.1
XP_016873976.1
4
888
XM_017018488.1
XP_016873977.1
4
888
XM_017018489.1
XP_016873978.1
4
849
XM_017018490.1
XP_016873979.1
4
875
XM_017018491.1
XP_016873980.1
4
844
XM_017018492.1
XP_016873981.1
4
888
XM_017018493.1
XP_016873982.1
4
875
XM_017018494.1
XP_016873983.1
4
888
XM_017018495.1
XP_016873984.1
4
888
XM_017018496.1
XP_016873985.1
4
844
XM_017018497.1
XP_016873986.1
4
849
XM_017018498.1
XP_016873987.1
4
888
XM_017018499.1
XP_016873988.1
4
857
XM_017018500.1
XP_016873989.1
4
888
XM_017018501.1
XP_016873990.1
4
844
XM_017018502.1
XP_016873991.1
4
857
XM_017018503.1
XP_016873992.1
4
844
XM_017018504.1
XP_016873993.1
4
857
XM_017018505.1
XP_016873994.1
4
844
XM_017018506.1
XP_016873995.1
4
849
XM_017018507.1
XP_016873996.1
4
875
XM_017018508.1
XP_016873997.1
4
888
XM_017018509.1
XP_016873998.1
4
844
XM_017018510.1
XP_016873999.1
4
888
GenBank mRNA
AB002356.1
-
4
860
AF440100.1
-
4
838
AF440101.1
-
4
838
AF440102.1
-
4
838
AF440103.1
-
4
838
AF440434.1
-
4
838
AK292442.1
-
4
828
BC040484.1
-
4
872
5271798
U44953.1
-
4
852
U77352.1
-
4
998
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
cell surface receptor signaling pathway
regulation of tumor necrosis factor-mediated signaling pathway
regulation of Rab protein signal transduction
regulation of apoptotic process
positive regulation of GTPase activity
regulation of cell cycle
execution phase of apoptosis
regulation of extrinsic apoptotic signaling pathway via death domain receptors
regulation of extrinsic apoptotic signaling pathway
death receptor binding
protein binding
Rab guanyl-nucleotide exchange factor activity
protein kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-whlt2:80/10.239.162.50:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0