Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MADD SIRNA Assays ›
Gene Symbol:
MADD
Gene Name:
MAP kinase activating death domain
Gene Aliases:
DENN, IG20, RAB3GEP
Chromosome Location:
Chr.11: 47269376 - 47330031 on Build GRCh38
UniGene ID:
Hs.82548
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139309
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MADD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135943.1
NP_001129415.1
4
1018
NM_001135944.1
NP_001129416.1
4
994
NM_003682.3
NP_003673.3
4
994
NM_130470.2
NP_569826.2
4
1160
NM_130471.2
NP_569827.2
4
994
NM_130472.2
NP_569828.2
4
994
NM_130473.2
NP_569829.2
4
994
NM_130474.2
NP_569830.2
4
994
NM_130475.2
NP_569831.1
4
1018
NM_130476.2
NP_569832.2
4
1018
XM_005253189.2
XP_005253246.1
4
1018
XM_005253196.2
XP_005253253.1
4
1018
XM_005253199.2
XP_005253256.1
4
1018
XM_005253200.2
XP_005253257.1
4
1018
XM_005253201.2
XP_005253258.1
4
1018
XM_005253203.2
XP_005253260.1
4
1018
XM_005253204.2
XP_005253261.1
4
1018
XM_005253205.1
XP_005253262.1
4
1018
XM_017018478.1
XP_016873967.1
4
1018
XM_017018479.1
XP_016873968.1
4
1018
XM_017018480.1
XP_016873969.1
4
1018
XM_017018481.1
XP_016873970.1
4
1018
XM_017018482.1
XP_016873971.1
4
1018
XM_017018483.1
XP_016873972.1
4
1018
XM_017018484.1
XP_016873973.1
4
1018
XM_017018485.1
XP_016873974.1
4
979
XM_017018486.1
XP_016873975.1
4
1018
XM_017018487.1
XP_016873976.1
4
1018
XM_017018488.1
XP_016873977.1
4
1018
XM_017018489.1
XP_016873978.1
4
979
XM_017018490.1
XP_016873979.1
4
1005
XM_017018491.1
XP_016873980.1
4
974
XM_017018492.1
XP_016873981.1
4
1018
XM_017018493.1
XP_016873982.1
4
1005
XM_017018494.1
XP_016873983.1
4
1018
XM_017018495.1
XP_016873984.1
4
1018
XM_017018496.1
XP_016873985.1
4
974
XM_017018497.1
XP_016873986.1
4
979
XM_017018498.1
XP_016873987.1
4
1018
XM_017018499.1
XP_016873988.1
4
987
XM_017018500.1
XP_016873989.1
4
1018
XM_017018501.1
XP_016873990.1
4
974
XM_017018502.1
XP_016873991.1
4
987
XM_017018503.1
XP_016873992.1
4
974
XM_017018504.1
XP_016873993.1
4
987
XM_017018505.1
XP_016873994.1
4
974
XM_017018506.1
XP_016873995.1
4
979
XM_017018507.1
XP_016873996.1
4
1005
XM_017018508.1
XP_016873997.1
4
1018
XM_017018509.1
XP_016873998.1
4
974
XM_017018510.1
XP_016873999.1
4
1018
GenBank mRNA
AB002356.1
-
4
990
AF440100.1
-
4
968
AF440101.1
-
4
968
AF440102.1
-
4
968
AF440103.1
-
4
968
AF440434.1
-
4
968
AK292442.1
-
4
958
BC040484.1
-
4
1002
5271798
U44953.1
-
4
982
U77352.1
-
4
1128
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
cell surface receptor signaling pathway
regulation of tumor necrosis factor-mediated signaling pathway
regulation of Rab protein signal transduction
regulation of apoptotic process
positive regulation of GTPase activity
regulation of cell cycle
execution phase of apoptosis
regulation of extrinsic apoptotic signaling pathway via death domain receptors
regulation of extrinsic apoptotic signaling pathway
death receptor binding
protein binding
Rab guanyl-nucleotide exchange factor activity
protein kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0