Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH7-TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037639.2 | 27 | 4621 | |||
NR_037640.2 | 27 | 4488 | ||||
NR_037641.2 | 26 | 4372 | ||||
GenBank mRNA | AK127008.1 | - | 11 | 2122 | ||
BX648088.1 | - | 27 | 4427 | |||
CR749830.1 | - | 27 | 4573 |
Gene Symbol: TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291333.1 | NP_001278262.1 | 4 | 530 | ||
NM_004623.4 | NP_004614.3 | 4 | 530 | |||
GenBank mRNA | AF063602.1 | - | 4 | 492 | ||
AF073887.1 | - | 4 | 476 | |||
AK025524.1 | - | 4 | 530 | |||
AK090964.1 | - | 4 | 610 | |||
AK222562.1 | - | 4 | 483 | |||
AK292426.1 | - | 4 | 491 | |||
BC001276.1 | - | 4 | 469 | 3462738 | ||
BT006817.1 | - | 4 | 443 |