Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025252.2 | NP_001020423.1 | 6 | 1579 | ||
NM_001025253.2 | NP_001020424.1 | 8 | 1366 | |||
NM_001287140.1 | NP_001274069.1 | 8 | 1648 | |||
NM_001287142.1 | NP_001274071.1 | 7 | 1621 | |||
NM_001287143.1 | NP_001274072.1 | 7 | 1606 | |||
NM_001287144.1 | NP_001274073.1 | 6 | 1290 | |||
NM_005079.3 | NP_005070.1 | 6 | 1297 | |||
NR_105033.1 | 9 | 1718 | ||||
NR_105034.1 | 5 | 1181 | ||||
NR_105035.1 | 7 | 1367 | ||||
NR_105036.1 | 8 | 1311 | ||||
NR_105037.1 | 7 | 1312 | ||||
GenBank mRNA | AF202897.1 | - | 6 | 1523 | ||
AK057075.1 | - | 8 | 1367 | |||
AK295468.1 | - | 8 | 1233 | |||
AK296615.1 | - | 6 | 1301 | |||
AK308983.1 | - | 8 | 1223 | |||
BC018117.1 | - | 6 | 1168 | 3916870 | ||
BX640835.1 | - | 6 | 1363 | |||
GQ499324.1 | - | 8 | 1633 | |||
GQ499325.1 | - | 7 | 1606 | |||
GQ499326.1 | - | 7 | 1591 | |||
GQ499327.1 | - | 6 | 1564 | |||
GQ499328.1 | - | 9 | 1703 | |||
S82081.1 | - | 6 | 1174 | |||
U18914.1 | - | 6 | 1172 |