Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270488.1 | NP_001257417.1 | 4 | 617 | ||
NM_005643.3 | NP_005634.1 | 5 | 714 | |||
XM_011514827.2 | XP_011513129.1 | 5 | 1104 | |||
GenBank mRNA | AF118094.1 | - | 5 | 454 | ||
AK302226.1 | - | 4 | 588 | |||
AK313475.1 | - | 5 | 655 | |||
BC021972.2 | - | 5 | 670 | 3935882 | ||
BT019404.1 | - | 5 | 600 | |||
BT019405.1 | - | 5 | 600 | |||
BX538195.1 | - | 2 | 1982 | |||
BX647568.1 | - | 3 | 2885 | |||
CR542068.1 | - | 5 | 600 | |||
D63705.1 | - | 5 | 683 | |||
X83928.1 | - | 5 | 692 |