Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291602.1 | NP_001278531.1 | 3 | 241 | ||
NM_006519.3 | NP_006510.1 | 4 | 283 | |||
GenBank mRNA | AK026669.1 | - | 2 | 2177 | ||
AK315601.1 | - | 4 | 240 | |||
BC029412.1 | - | 4 | 236 | 4074224 | ||
BC105588.1 | - | 4 | 232 | 6723966 | ||
CR456931.1 | - | 4 | 205 | |||
D50663.1 | - | 4 | 217 | |||
EU862237.1 | - | 4 | 209 | |||
HY017024.1 | - | 3 | 194 | |||
HY171700.1 | - | 4 | 286 | |||
U56255.1 | - | 4 | 205 |