Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291602.1 | NP_001278531.1 | 4 | 346 | ||
NM_001291603.1 | NP_001278532.1 | 5 | 368 | |||
NM_006519.3 | NP_006510.1 | 5 | 388 | |||
GenBank mRNA | AK026669.1 | - | 2 | 2512 | ||
AK315601.1 | - | 5 | 345 | |||
BC029412.1 | - | 5 | 341 | 4074224 | ||
BC105588.1 | - | 5 | 337 | 6723966 | ||
CR456931.1 | - | 5 | 310 | |||
D50663.1 | - | 5 | 322 | |||
DN915418.1 | - | 4 | 282 | |||
EU862237.1 | - | 5 | 314 | |||
HY017024.1 | - | 4 | 299 | |||
HY171700.1 | - | 5 | 391 | |||
U56255.1 | - | 5 | 310 |