Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006767.3 | NP_006758.2 | 14 | 1700 | ||
GenBank mRNA | AK296347.1 | - | 13 | 1581 | ||
AK310747.1 | - | 12 | 1303 | |||
AK313502.1 | - | 9 | 1102 | |||
BC026214.1 | - | 14 | 1602 | 4399716 | ||
CT841521.1 | - | 14 | 1616 | |||
D38496.1 | - | 14 | 1595 |