Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006950.3 | NP_008881.2 | 9 | 1271 | ||
NM_133499.2 | NP_598006.1 | 9 | 1271 | |||
GenBank mRNA | AL833961.1 | - | 1 | 94 |