Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256647.1 | NP_001243576.1 | 5 | 662 | ||
NM_007121.5 | NP_009052.3 | 6 | 953 | |||
GenBank mRNA | AB307697.1 | - | 4 | 672 | ||
AK091535.1 | - | 10 | 1424 | |||
AK297978.1 | - | 5 | 627 | |||
BC007790.1 | - | 6 | 820 | 3636961 | ||
BC033500.1 | - | 6 | 871 | 5178510 | ||
BC047750.2 | - | 6 | 907 | 6469687 | ||
BC074500.1 | - | 6 | 888 | 5211737 | ||
CR456706.1 | - | 4 | 672 | |||
CR749648.1 | - | 6 | 903 | |||
HQ692835.1 | - | 4 | 672 | |||
U07132.1 | - | 6 | 916 | |||
U14534.1 | - | 4 | 654 |