Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18RAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003853.3 | NP_003844.1 | 11 | 1714 | ||
XM_011512087.2 | XP_011510389.1 | 11 | 1488 | |||
XM_011512088.2 | XP_011510390.1 | 8 | 936 | |||
XM_017005173.1 | XP_016860662.1 | 4 | 1069 | |||
GenBank mRNA | AF077346.1 | - | 11 | 1709 | ||
BC069630.1 | - | 9 | 1311 | 7262472 | ||
BC106764.2 | - | 9 | 1113 | 40026248 | ||
BC106765.2 | - | 10 | 1438 | 40026249 | ||
BC137474.1 | - | 9 | 1254 | 9021474 | ||
BC137475.1 | - | 9 | 1254 | 9021475 |