Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093728.2 | NP_001087197.1 | 6, 7 | 936 | ||
NM_001261817.1 | NP_001248746.1 | 6, 7 | 921 | |||
NM_001261818.1 | NP_001248747.1 | 5, 6 | 798 | |||
NM_001261819.1 | NP_001248748.1 | 5, 6 | 783 | |||
NM_001261820.1 | NP_001248749.1 | 7, 8 | 1142 | |||
NM_001261821.1 | NP_001248750.1 | 7, 8 | 1127 | |||
NM_018679.5 | NP_061149.1 | 6, 7 | 1030 | |||
XM_005249339.2 | XP_005249396.1 | 5, 6 | 691 | |||
XM_011514830.1 | XP_011513132.1 | 6, 7 | 1168 | |||
XM_011514831.1 | XP_011513133.1 | 4, 5 | 604 | |||
XM_011514832.1 | XP_011513134.1 | 3, 4 | 468 | |||
GenBank mRNA | AF260330.1 | - | 3, 4 | 479 | ||
AF269223.1 | - | 5, 6 | 714 | |||
AF536532.1 | - | 7, 8 | 982 | |||
AF536533.1 | - | 6, 7 | 969 | |||
AK057234.1 | - | 6, 7 | 753 | |||
AK098590.1 | - | 4, 5 | 525 | |||
AK301778.1 | - | 6, 7 | 812 | |||
AK301975.1 | - | 5, 6 | 719 | |||
AK302022.1 | - | 5, 6 | 684 | |||
AK302375.1 | - | 6, 7 | 861 | |||
AK302456.1 | - | 6, 7 | 842 | |||
AK315078.1 | - | 6, 7 | 963 | |||
AK316403.1 | - | 6, 7 | 745 | |||
AY069943.1 | - | 5, 6 | 738 | |||
BC033729.1 | - | 6, 7 | 882 | 5167179 | ||
BC040003.1 | - | 7, 8 | 984 | 5741923 | ||
BC048418.1 | - | 6, 7 | 960 | 5298037 | ||
BC090050.1 | - | 7, 8 | 988 | 30387768 |