Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021138.3 | NP_066961.2 | 7 | 680 | ||
XM_011518973.2 | XP_011517275.1 | 8 | 984 | |||
XM_011518974.2 | XP_011517276.1 | 7 | 1585 | |||
XM_011518975.2 | XP_011517277.1 | 8 | 912 | |||
XM_011518976.2 | XP_011517278.1 | 8 | 953 | |||
XM_011518977.2 | XP_011517279.1 | 7 | 673 | |||
XM_011518978.2 | XP_011517280.1 | 7 | 2094 | |||
XM_017015094.1 | XP_016870583.1 | 7 | 728 | |||
XM_017015095.1 | XP_016870584.1 | 7 | 1438 | |||
GenBank mRNA | AK289722.1 | - | 8 | 796 | ||
AK294607.1 | - | 7 | 539 | |||
AK298370.1 | - | 7 | 644 | |||
BC032410.1 | - | 6 | 666 | 5216790 | ||
BC033810.1 | - | 7 | 668 | 5182942 | ||
BC043492.1 | - | 7 | 680 | 5767794 | ||
BC064662.1 | - | 6 | 675 | 6528158 | ||
BX538160.1 | - | 6 | 622 | |||
HM991672.1 | - | 6 | 623 |