Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SVIL SIRNA Assays ›
Gene Symbol:
SVIL
Gene Name:
supervillin
Gene Aliases:
-
Chromosome Location:
Chr.10: 29457338 - 29737001 on Build GRCh38
UniGene ID:
Hs.499209
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139866
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SVIL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001323599.1
NP_001310528.1
16
2747
NM_001323600.1
NP_001310529.1
14
2348
NM_003174.3
NP_003165.2
13
2133
NM_021738.2
NP_068506.2
15
3714
XM_005252570.2
XP_005252627.1
17
3611
XM_005252571.3
XP_005252628.1
17
3537
XM_005252573.2
XP_005252630.1
15
3743
XM_011519630.2
XP_011517932.2
15
4456
XM_011519631.2
XP_011517933.2
14
4360
XM_011519632.2
XP_011517934.2
14
4285
XM_011519633.2
XP_011517935.2
14
4360
XM_011519634.1
XP_011517936.1
17
3669
XM_011519635.1
XP_011517937.1
15
3636
XM_011519636.2
XP_011517938.2
14
3526
XM_011519637.2
XP_011517939.2
12
3274
XM_011519638.2
XP_011517940.2
11
3178
XM_017016575.1
XP_016872064.1
15
4456
XM_017016576.1
XP_016872065.1
14
4375
XM_017016577.1
XP_016872066.1
15
4456
XM_017016578.1
XP_016872067.1
13
3430
XM_017016579.1
XP_016872068.1
14
3526
XM_017016580.1
XP_016872069.1
13
3355
XM_017016581.1
XP_016872070.1
12
3274
XM_017016582.1
XP_016872071.1
11
3178
XM_017016583.1
XP_016872072.1
15
4456
GenBank mRNA
AF051850.1
-
13
2133
AF051851.1
-
9
1783
AF109135.1
-
15
3714
JX467682.1
-
10
2865
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
skeletal muscle tissue development
positive regulation of cytokinesis
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0