Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001056.3 | NP_001047.1 | 8 | 1994 | ||
NM_176825.2 | NP_789795.1 | 9 | 2027 | |||
XM_005264013.3 | XP_005264070.1 | 8 | 1815 | |||
GenBank mRNA | AB008164.1 | - | 8 | 1664 | ||
BM849361.1 | - | 1 | 317 | |||
R86241.1 | - | 1 | 142 |