Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007123.5 | NP_009054.5 | 2 | 556 | ||
NM_206933.2 | NP_996816.2 | 2 | 556 | |||
GenBank mRNA | AF055580.2 | - | 2 | 556 | ||
AY481573.1 | - | 2 | 556 |