Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143906.1 | NP_001137378.1 | 5 | 1241 | ||
NM_006700.2 | NP_006691.1 | 5 | 710 | |||
GenBank mRNA | AB007447.1 | - | 5 | 678 | ||
AK122620.1 | - | 5 | 1226 | |||
AK291331.1 | - | 5 | 695 | |||
AK309916.1 | - | 5 | 717 | |||
AK312601.1 | - | 5 | 710 | |||
BC003553.2 | - | 4 | 636 | 3547039 |