Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143906.1 | NP_001137378.1 | 10 | 1985 | ||
NM_006700.2 | NP_006691.1 | 10 | 1454 | |||
GenBank mRNA | AB007447.1 | - | 10 | 1422 | ||
AK122620.1 | - | 10 | 1969 | |||
AK291331.1 | - | 10 | 1439 | |||
AK312601.1 | - | 10 | 1454 | |||
BC003553.2 | - | 9 | 1380 | 3547039 |