Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272071.1 | NP_001259000.1 | 6 | 1068 | ||
NM_003916.4 | NP_003907.3 | 5 | 1059 | |||
GenBank mRNA | AF087876.1 | - | 4 | 850 | ||
AF091077.1 | - | 4 | 737 | |||
AF251295.1 | - | 5 | 817 | |||
AK125525.1 | - | 1 | 2212 | |||
AK130075.1 | - | 5 | 862 | |||
BC001117.2 | - | 5 | 806 | 3050001 | ||
BC071867.1 | - | 5 | 807 | 4556064 | ||
BX537780.1 | - | 5 | 756 |