Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144831.1 | NP_001138303.1 | 4 | 566 | ||
NM_001267700.1 | NP_001254629.1 | 4 | 566 | |||
GenBank mRNA | AF126021.1 | - | 4 | 541 | ||
AF150962.1 | - | 4 | 479 | |||
AF178980.1 | - | 4 | 455 | |||
AK298217.1 | - | 4 | 486 | |||
AK301316.1 | - | 4 | 530 | |||
BC013228.1 | - | 4 | 449 | |||
BC014766.1 | - | 4 | 436 | 3452360 | ||
BC110322.1 | - | 4 | 566 | 5540527 | ||
U72511.1 | - | 2 | 94 |