Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142633.2 | NP_001136105.1 | 19 | 3030 | ||
NM_001251851.1 | NP_001238780.1 | 19 | 3092 | |||
NM_001251852.1 | NP_001238781.1 | 18 | 2914 | |||
NM_001251853.1 | NP_001238782.1 | 18 | 2876 | |||
NM_001251855.1 | NP_001238784.1 | 18 | 2855 | |||
NM_014308.3 | NP_055123.2 | 19 | 2967 | |||
XM_005256580.3 | XP_005256637.1 | 19 | 3022 | |||
GenBank mRNA | AF128881.1 | - | 19 | 2967 | ||
BC028212.1 | - | 18 | 2846 | 5215094 |