Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286414.2 | NP_001273343.1 | 13 | 1607 | ||
NM_014444.4 | NP_055259.2 | 13 | 1604 | |||
XM_011521454.2 | XP_011519756.1 | 13 | 1607 | |||
XM_011521455.2 | XP_011519757.1 | 13 | 1505 | |||
XM_017022078.1 | XP_016877567.1 | 13 | 1604 | |||
GenBank mRNA | AJ249677.1 | - | 13 | 1313 | ||
AK001639.1 | - | 9 | 918 | |||
AK026551.1 | - | 13 | 1594 | |||
AK027703.1 | - | 13 | 1565 | |||
AK127306.1 | - | 3 | 1748 | |||
BC000966.2 | - | 11 | 1067 | 3446803 | ||
BC009870.2 | - | 13 | 1529 | 3940649 | ||
BC012801.2 | - | 13 | 1548 | 2821891 |