Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303627.1 | NP_001290556.1 | 5 | 1859 | ||
NM_001303628.1 | NP_001290557.1 | 3 | 1907 | |||
NM_014730.3 | NP_055545.1 | 5 | 2142 | |||
XM_011539031.1 | XP_011537333.1 | 5 | 6557 | |||
XM_011539032.1 | XP_011537334.1 | 6 | 6245 | |||
GenBank mRNA | BF794298.1 | - | 42 | |||
CR936818.1 | - | 2 | 1311 | |||
D63486.1 | - | 5 | 2099 | |||
U88835.1 | - | 1 | 615 |