Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293640.1 | NP_001280569.1 | 7 | 575 | ||
NM_015275.2 | NP_056090.1 | 7 | 575 | |||
XM_011538073.2 | XP_011536375.1 | 7 | 575 | |||
XM_011538074.2 | XP_011536376.1 | 7 | 529 | |||
XM_011538077.2 | XP_011536379.1 | 7 | 575 | |||
XM_017019082.1 | XP_016874571.1 | 7 | 575 | |||
GenBank mRNA | AB028956.1 | - | 7 | 557 | ||
AK001657.1 | - | 7 | 575 | |||
AK001728.1 | - | 7 | 540 | |||
AK023146.1 | - | 4 | 210 | |||
AL137753.1 | - | 4 | 186 | |||
BC104992.1 | - | 7 | 517 | 8143995 | ||
BC104994.1 | - | 7 | 517 | 8143997 | ||
BC110850.1 | - | 7 | 526 | |||
BC143373.1 | - | 7 | 517 | 9051878 |