Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015433.2 | NP_056248.2 | 3 | 2227 | ||
NM_206914.1 | NP_996797.1 | 4 | 2366 | |||
GenBank mRNA | AF455816.1 | - | 3 | 2227 | ||
AK024983.1 | - | 4 | 2255 | |||
AL050100.1 | - | 4 | 2287 | |||
BC016395.1 | - | 3 | 2227 | 3882414 |