Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016626.4 | NP_057710.3 | 2 | 3530 | ||
GenBank mRNA | AF208855.1 | - | 1 | 1647 | ||
AK096190.1 | - | 2 | 2771 | |||
AL833519.1 | - | 1 | 2390 | |||
BC041122.1 | - | 1 | 1821 |