Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184856.1 | NP_001171785.1 | 2, 3 | 297 | ||
NM_017566.3 | NP_060036.2 | 2, 3 | 297 | |||
XM_005255994.3 | XP_005256051.1 | 2, 3 | 302 | |||
XM_006721201.1 | XP_006721264.1 | 2, 3 | 302 | |||
XM_006721202.1 | XP_006721265.1 | 2, 3 | 302 | |||
XM_006721203.2 | XP_006721266.1 | 2, 3 | 302 | |||
XM_006721204.3 | XP_006721267.1 | 2, 3 | 302 | |||
XM_017023344.1 | XP_016878833.1 | 2, 3 | 302 | |||
XM_017023345.1 | XP_016878834.1 | 2, 3 | 237 | |||
GenBank mRNA | AK023986.1 | - | 2, 3 | 254 | ||
BC011680.2 | - | 2, 3 | 281 | 4303424 | ||
BC022969.1 | - | 2, 3 | 241 | 5112326 | ||
BP215584.1 | - | 2, 3 | 297 |