Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VEZT SIRNA Assays ›
Gene Symbol:
VEZT
Gene Name:
vezatin, adherens junctions transmembrane protein
Gene Aliases:
VEZATIN
Chromosome Location:
Chr.12: 95217746 - 95302799 on Build GRCh38
UniGene ID:
Hs.24135
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140403
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VEZT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017599.3
NP_060069.3
4
443
NR_038241.1
5
516
NR_038242.1
4
439
XM_006719477.3
XP_006719540.1
4
443
XM_006719478.2
XP_006719541.1
3
337
XM_006719479.2
XP_006719542.1
5
514
XM_006719480.3
XP_006719543.1
5
491
XM_006719481.2
XP_006719544.1
4
361
XM_006719482.3
XP_006719545.1
4
742
XM_006719483.3
XP_006719546.1
4
759
XM_006719484.3
XP_006719547.1
3
669
XM_006719490.3
XP_006719553.1
5
514
XM_011538543.1
XP_011536845.1
5
436
XM_011538544.1
XP_011536846.1
5
436
XM_011538545.2
XP_011536847.1
4
404
XM_011538546.1
XP_011536848.1
6
507
XM_011538547.1
XP_011536849.1
6
551
XM_011538549.1
XP_011536851.1
5
436
XM_011538550.2
XP_011536852.1
5
436
XM_011538551.2
XP_011536853.1
5
436
XM_011538552.1
XP_011536854.1
5
510
XM_011538553.2
XP_011536855.1
5
440
XM_017019586.1
XP_016875075.1
4
404
XM_017019587.1
XP_016875076.1
3
337
XM_017019588.1
XP_016875077.1
6
507
XM_017019589.1
XP_016875078.1
5
491
XM_017019590.1
XP_016875079.1
6
551
XM_017019591.1
XP_016875080.1
4
742
XM_017019593.1
XP_016875082.1
6
559
XM_017019594.1
XP_016875083.1
5
421
XM_017019595.1
XP_016875084.1
4
443
XM_017019596.1
XP_016875085.1
4
443
XM_017019597.1
XP_016875086.1
4
443
XM_017019598.1
XP_016875087.1
4
443
XM_017019599.1
XP_016875088.1
4
404
XM_017019600.1
XP_016875089.1
4
759
XM_017019601.1
XP_016875090.1
5
510
XM_017019602.1
XP_016875091.1
6
507
XM_017019603.1
XP_016875092.1
4
759
GenBank mRNA
AF216644.1
-
4
370
AF225417.1
-
4
414
AK225303.1
-
5
456
AK225356.1
-
5
453
AK307649.1
-
5
510
AK309943.1
-
4
378
AL137497.1
-
5
438
BC028597.1
-
7
764
BC064939.1
-
5
433
6159768
DB136255.1
-
4
443
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
single organismal cell-cell adhesion
myosin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0