Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271762.1 | NP_001258691.1 | 3 | 572 | ||
NM_001271763.1 | NP_001258692.1 | 4 | 652 | |||
NM_017803.4 | NP_060273.1 | 4 | 652 | |||
GenBank mRNA | AB101210.1 | - | 4 | 316 | ||
AK000406.1 | - | 3 | 219 | |||
AK290578.1 | - | 4 | 354 | |||
AK296663.1 | - | 4 | 304 | |||
BC006527.2 | - | 4 | 295 | 2988128 |