Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017822.3 | NP_060292.3 | 5, 6 | 786 | ||
GenBank mRNA | AK000443.1 | - | 4, 5 | 630 | ||
AK022732.1 | - | 4, 5 | 524 | |||
AK094528.1 | - | 4, 5 | 1309 | |||
AL834467.1 | - | 2, 3 | 205 | |||
BC009746.2 | - | 5, 6 | 754 | 3503970 | ||
BC013900.2 | - | 4, 5 | 706 | 3928141 |