Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BANP SIRNA Assays ›
Gene Symbol:
BANP
Gene Name:
BTG3 associated nuclear protein
Gene Aliases:
BEND1, SMAR1, SMARBP1
Chromosome Location:
Chr.16: 87949216 - 88077318 on Build GRCh38
UniGene ID:
Hs.461705
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140457
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BANP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173539.1
NP_001167010.1
4
504
NM_001173540.1
NP_001167011.1
4
522
NM_001173541.1
NP_001167012.1
4
504
NM_001173542.1
NP_001167013.1
4
543
NM_001173543.1
NP_001167014.1
4
549
NM_017869.3
NP_060339.2
4
504
NM_079837.2
NP_524576.2
4
504
XM_005256008.3
XP_005256065.1
4
520
XM_005256009.3
XP_005256066.1
4
502
XM_011523170.2
XP_011521472.1
4
520
XM_011523172.2
XP_011521474.1
5
765
XM_011523174.2
XP_011521476.1
4
1291
XM_011523175.2
XP_011521477.1
4
922
XM_011523176.2
XP_011521478.1
4
984
XM_011523177.2
XP_011521479.1
4
1492
XM_011523178.2
XP_011521480.1
4
520
XM_011523179.2
XP_011521481.1
4
502
XM_011523180.2
XP_011521482.1
5
584
XM_011523181.2
XP_011521483.1
4
520
XM_011523182.1
XP_011521484.1
4
520
XM_011523183.2
XP_011521485.1
4
497
XM_011523184.2
XP_011521486.1
4
453
XM_011523186.2
XP_011521488.1
2
282
XM_017023378.1
XP_016878867.1
5
624
XM_017023379.1
XP_016878868.1
4
904
XM_017023380.1
XP_016878869.1
4
504
XM_017023381.1
XP_016878870.1
4
522
XM_017023383.1
XP_016878872.1
4
469
GenBank mRNA
AK094158.1
-
4
550
AK125232.1
-
4
502
AK293472.1
-
4
466
AK297945.1
-
4
476
AK315089.1
-
4
502
AK315535.1
-
4
543
BC009424.1
-
4
449
3530821
DA632793.1
-
4
504
KU178664.1
-
3
352
KU178665.1
-
3
352
KU178666.1
-
3
352
KU178667.1
-
3
352
KU178668.1
-
3
352
KU178669.1
-
3
352
KU178670.1
-
3
374
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
cell cycle
multicellular organism development
covalent chromatin modification
protein localization to nucleus
negative regulation of protein catabolic process
positive regulation of transcription, DNA-templated
regulation of signal transduction by p53 class mediator
p53 binding
DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0