Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320772.1 | NP_001307701.1 | 3, 4 | 458 | ||
NM_017983.6 | NP_060453.3 | 4, 5 | 541 | |||
NR_135470.1 | 4, 5 | 541 | ||||
NR_135471.1 | 4, 5 | 541 | ||||
XM_017024808.1 | XP_016880297.1 | 2, 3 | 2495 | |||
GenBank mRNA | AK000917.1 | - | 3, 4 | 459 | ||
AK301830.1 | - | 3, 4 | 404 | |||
AY691424.1 | - | 4, 5 | 438 | |||
BC039867.2 | - | 4, 5 | 485 | 6055425 | ||
BI764142.1 | - | 4, 5 | 530 |