Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 4 | 466 | ||
NM_018427.4 | NP_060897.3 | 5 | 556 | |||
XM_005255376.1 | XP_005255433.1 | 5 | 465 | |||
XM_005255377.4 | XP_005255434.1 | 4 | 414 | |||
XM_011522541.1 | XP_011520843.1 | 5 | 465 | |||
GenBank mRNA | AF227156.1 | - | 5 | 522 | ||
AJ272050.1 | - | 5 | 476 | |||
AK225662.1 | - | 5 | 532 | |||
AK295486.1 | - | 2 | 149 | |||
AK300253.1 | - | 4 | 375 | |||
AK302994.1 | - | 2 | 203 | |||
AK303405.1 | - | 2 | 148 | |||
AK304856.1 | - | 5 | 422 | |||
AK314769.1 | - | 5 | 470 | |||
AK316520.1 | - | 4 | 380 | |||
BC036182.1 | - | 5 | 471 | |||
BC104660.1 | - | 5 | 478 | |||
BC132688.1 | - | 5 | 473 | 40146710 | ||
BC132690.1 | - | 5 | 473 | 40146712 |
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003370.2 | 5 | 431 | |||
GenBank mRNA | BC009198.2 | - | 6 | 661 | ||
BC068999.1 | - | 6 | 661 | |||
BX101439.1 | - | 5 | 521 | |||
CN397262.1 | - | 5 | 438 |
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 5 | 645 | |||
GenBank mRNA | AK225670.1 | - | 5 | 493 | ||
AK298442.1 | - | 4 | 378 | |||
AK301253.1 | - | 5 | 465 |