Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288732.1 | NP_001275661.1 | 2 | 556 | ||
NM_001288733.1 | NP_001275662.1 | 2 | 548 | |||
NM_018469.4 | NP_060939.3 | 2 | 556 | |||
XM_011524998.1 | XP_011523300.1 | 2 | 545 | |||
XM_011524999.1 | XP_011523301.1 | 2 | 469 | |||
XM_011525000.1 | XP_011523302.1 | 2 | 526 | |||
XM_017024846.1 | XP_016880335.1 | 2 | 526 | |||
XM_017024847.1 | XP_016880336.1 | 2 | 526 | |||
GenBank mRNA | AB051525.2 | - | 1 | 375 | ||
AK091214.1 | - | 2 | 474 | |||
AL832371.1 | - | 2 | 456 | |||
AL834251.2 | - | 2 | 467 | |||
BC036672.1 | - | 2 | 527 | |||
BC040521.1 | - | 2 | 460 | 4837394 | ||
BC040522.1 | - | 2 | 529 | |||
CR627433.1 | - | 2 | 549 |