Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019116.2 | NP_061989.2 | 7 | 1284 | ||
XM_011545894.2 | XP_011544196.1 | 6 | 1207 | |||
GenBank mRNA | AK092696.1 | - | 7 | 1098 | ||
AK093029.1 | - | 2 | 2368 | |||
AK096189.1 | - | 3 | 365 | |||
AK124139.1 | - | 8 | 1060 |