Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019613.3 | NP_062559.2 | 5 | 602 | ||
XM_005256377.4 | XP_005256434.1 | 4 | 425 | |||
XM_017024862.1 | XP_016880351.1 | 6 | 1939 | |||
XM_017024863.1 | XP_016880352.1 | 7 | 737 | |||
GenBank mRNA | AF091083.1 | - | 3 | 245 | ||
AK297477.1 | - | 5 | 356 | |||
AM182326.1 | - | 5 | 407 | |||
AY691427.1 | - | 3 | 233 | |||
BC000974.2 | - | 5 | 385 | |||
BC007838.1 | - | 5 | 574 | |||
CR456770.1 | - | 3 | 233 |