Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031628.1 | NP_001026798.1 | 4 | 1041 | ||
NM_001033873.1 | NP_001029045.1 | 4 | 1030 | |||
GenBank mRNA | AF091087.1 | - | 1 | 720 | ||
AK001839.1 | - | 4 | 1004 | |||
BC003379.1 | - | 1 | 716 | |||
BC008712.1 | - | 4 | 1018 | 3546069 | ||
BC045658.1 | - | 4 | 1513 |