Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTHFSD SIRNA Assays ›
Gene Symbol:
MTHFSD
Gene Name:
methenyltetrahydrofolate synthetase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.16: 86530176 - 86555235 on Build GRCh38
UniGene ID:
Hs.343627
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140723
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTHFSD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159377.1
NP_001152849.1
8
975
NM_001159378.1
NP_001152850.1
8
975
NM_001159379.1
NP_001152851.1
8
972
NM_001159380.1
NP_001152852.1
8
939
NM_022764.2
NP_073601.2
8
972
NR_027489.1
9
1045
NR_027490.1
8
1035
XM_005256101.1
XP_005256158.1
8
1015
XM_005256105.1
XP_005256162.1
7
930
XM_005256106.1
XP_005256163.1
7
839
XM_006721247.3
XP_006721310.1
7
846
XM_011523280.2
XP_011521582.1
8
1015
XM_011523285.1
XP_011521587.1
9
1013
XM_011523286.1
XP_011521588.1
9
1113
XM_011523287.1
XP_011521589.1
9
1017
XM_011523288.1
XP_011521590.1
8
945
XM_011523289.1
XP_011521591.1
8
1022
XM_017023571.1
XP_016879060.1
7
861
XM_017023572.1
XP_016879061.1
7
858
XM_017023573.1
XP_016879062.1
7
838
XM_017023574.1
XP_016879063.1
8
938
GenBank mRNA
AK023060.1
-
8
950
AK023955.1
-
8
929
AK096836.1
-
9
1045
AK125015.1
-
1
1563
AK297726.1
-
8
972
BC112057.1
-
8
955
8327525
BC112059.1
-
8
955
8327527
BC143702.1
-
8
940
9052216
BC143704.1
-
8
940
BC143705.1
-
8
939
BC143707.1
-
8
943
9052221
Back To Top
More Information
Panther Classification:
Molecular Function -
ligase
Gene Ontology Categories:
Process(es)
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0