Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286191.1 | NP_001273120.1 | 4 | 462 | ||
NM_001286192.1 | NP_001273121.1 | 4 | 369 | |||
NM_001286195.1 | NP_001273124.1 | 4 | 369 | |||
NM_001286196.1 | NP_001273125.1 | 4 | 369 | |||
NM_001286197.1 | NP_001273126.1 | 3 | 270 | |||
NM_001286198.1 | NP_001273127.1 | 5 | 626 | |||
NM_022895.2 | NP_075046.1 | 4 | 369 | |||
NR_104409.1 | 4 | 379 | ||||
XM_017019829.1 | XP_016875318.1 | 4 | 453 | |||
GenBank mRNA | AK022510.1 | - | 4 | 364 | ||
AK222630.1 | - | 4 | 354 | |||
AK225162.1 | - | 4 | 364 | |||
AK299170.1 | - | 5 | 612 | |||
AK301566.1 | - | 4 | 354 | |||
AK307971.1 | - | 4 | 464 | |||
BC014661.1 | - | 4 | 343 | 4860511 | ||
BP201132.1 | - | 4 | 349 | |||
BP343829.1 | - | 3 | 197 | |||
DA014862.1 | - | 4 | 368 | |||
DC297725.1 | - | 4 | 356 |