Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024042.3 | NP_076947.1 | 3, 4 | 679 | ||
GenBank mRNA | BC000662.2 | - | 3, 4 | 679 | 3347703 |