Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MFSD11 SIRNA Assays ›
Gene Symbol:
MFSD11
Gene Name:
major facilitator superfamily domain containing 11
Gene Aliases:
ET
Chromosome Location:
Chr.17: 76736565 - 76803805 on Build GRCh38
UniGene ID:
Hs.732019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140787
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MFSD11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242532.1
NP_001229461.1
2
970
NM_001242533.1
NP_001229462.1
3
854
NM_001242534.1
NP_001229463.1
3
716
NM_001242535.1
NP_001229464.1
3
665
NM_001242536.1
NP_001229465.1
3
854
NM_001242537.1
NP_001229466.1
3
665
NM_024311.3
NP_077287.1
3
572
XM_011525236.2
XP_011523538.1
2
234
XM_011525237.2
XP_011523539.2
3
677
XM_011525238.2
XP_011523540.2
3
277
XM_011525239.2
XP_011523541.2
3
247
XM_011525240.2
XP_011523542.2
3
273
XM_011525241.2
XP_011523543.2
3
283
XM_011525242.1
XP_011523544.1
2
234
XM_011525244.1
XP_011523546.1
2
234
XM_011525247.2
XP_011523549.2
3
677
XM_017025065.1
XP_016880554.1
3
305
XM_017025066.1
XP_016880555.1
3
247
XM_017025067.1
XP_016880556.1
3
305
XM_017025068.1
XP_016880557.1
3
677
XM_017025069.1
XP_016880558.1
2
464
XM_017025070.1
XP_016880559.1
2
464
GenBank mRNA
AF015184.1
-
3
284
AF015185.1
-
3
471
AF015186.1
-
3
428
AF015190.1
-
3
716
AK000233.1
-
2
352
AK025849.1
-
2
107
AK308058.1
-
3
309
BC002753.2
-
3
372
3629029
BC062563.1
-
3
465
5140496
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0