Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPTAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024312.4 | NP_077288.2 | 5 | 725 | ||
XM_006719593.2 | XP_006719656.1 | 5 | 720 | |||
XM_011538731.2 | XP_011537033.1 | 5 | 721 | |||
XM_017019961.1 | XP_016875450.1 | 5 | 622 | |||
XM_017019962.1 | XP_016875451.1 | 5 | 704 | |||
GenBank mRNA | AK056137.1 | - | 5 | 596 | ||
AL832172.1 | - | 1 | 167 | |||
AM085438.1 | - | 5 | 712 | |||
AY687932.1 | - | 5 | 697 | |||
BC071687.1 | - | 5 | 673 | |||
BC131787.1 | - | 5 | 653 | 40126580 |