Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142350.1 | NP_001135822.1 | 9, 10 | 925 | ||
NM_024339.3 | NP_077315.2 | 9, 10 | 925 | |||
GenBank mRNA | AK075330.1 | - | 9, 10 | 892 | ||
AK314086.1 | - | 9, 10 | 903 | |||
AL832548.1 | - | 8, 9 | 529 | |||
BC003118.1 | - | 10, 11 | 805 | 3537243 | ||
BC050674.1 | - | 9, 10 | 864 | 6067702 |