Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193522.1 | NP_001180451.1 | 7 | 564 | ||
NM_001193523.1 | NP_001180452.1 | 7 | 677 | |||
NM_001193524.1 | NP_001180453.1 | 7 | 590 | |||
NM_024519.3 | NP_078795.2 | 7 | 555 | |||
XM_011523321.1 | XP_011521623.1 | 7 | 1342 | |||
XM_011523322.1 | XP_011521624.1 | 7 | 537 | |||
XM_011523324.2 | XP_011521626.1 | 7 | 597 | |||
XM_011523325.1 | XP_011521627.1 | 7 | 552 | |||
XM_011523326.1 | XP_011521628.1 | 7 | 751 | |||
XM_017023663.1 | XP_016879152.1 | 7 | 537 | |||
XM_017023664.1 | XP_016879153.1 | 6 | 450 | |||
GenBank mRNA | AB067517.1 | - | 6 | 414 | ||
AK023787.1 | - | 2 | 47 | |||
AK097087.1 | - | 6 | 805 | |||
AK127792.1 | - | 7 | 564 | |||
AK293748.1 | - | 7 | 587 | |||
AK295677.1 | - | 7 | 521 | |||
AK307467.1 | - | 7 | 508 | |||
CN401995.1 | - | 7 | 555 |