Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024816.2 | NP_079092.2 | 8 | 1190 | ||
GenBank mRNA | AK026935.1 | - | 8 | 1190 | ||
AK092299.1 | - | 8 | 1731 | |||
AK295229.1 | - | 7 | 962 | |||
BC046118.1 | - | 8 | 1184 | |||
BC058900.1 | - | 7 | 1097 | 5415624 |