Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270526.1 | NP_001257455.1 | 3, 4 | 357 | ||
NM_001270527.1 | NP_001257456.1 | 3, 4 | 357 | |||
NM_004099.5 | NP_004090.4 | 3, 4 | 357 | |||
NR_073037.1 | 4, 5 | 487 | ||||
GenBank mRNA | AK302977.1 | - | 3, 4 | 283 | ||
AK304449.1 | - | 4, 5 | 431 | |||
AK304518.1 | - | 3, 4 | 301 | |||
AK307999.1 | - | 3, 4 | 301 | |||
BC010703.1 | - | 3, 4 | 279 | 3873494 | ||
BI553227.1 | - | 3, 4 | 286 | |||
BM451470.1 | - | 321 | ||||
BP208523.1 | - | 3, 4 | 284 | |||
CR542100.1 | - | 3, 4 | 240 | |||
DC368197.1 | - | 3, 4 | 357 | |||
M81635.1 | - | 3, 4 | 261 | |||
X60067.1 | - | 3, 4 | 240 |