Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195059.1 | NP_001181988.1 | 3, 4 | 364 | ||
NM_025201.4 | NP_079477.2 | 4, 5 | 514 | |||
GenBank mRNA | AF318373.1 | - | 1, 2 | 112 | ||
AF494534.1 | - | 4, 5 | 463 | |||
AK096203.1 | - | 4, 5 | 482 | |||
BC008744.2 | - | 3, 4 | 325 | 3346334 |