Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3L-AARSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136042.2 | NP_001129514.2 | 8 | 1056 | ||
NM_025267.3 | NP_079543.1 | 8 | 672 | |||
GenBank mRNA | AF039235.1 | - | 8 | 761 | ||
AK094084.1 | - | 8 | 610 | |||
AK295447.1 | - | 8 | 1056 | |||
BC004172.2 | - | 8 | 672 | 2823004 | ||
BC019324.1 | - | 8 | 672 | 2823004 |
Gene Symbol: AARSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261434.1 | NP_001248363.1 | 3 | 246 | ||
GenBank mRNA | AK309477.1 | - | 3 | 193 | ||
AL515977.3 | - | 3 | 191 | |||
BP368900.1 | - | 3 | 224 | |||
CA951145.1 | - | 3 | 215 |