Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243650.1 | NP_001230579.1 | 2 | 467 | ||
NM_030819.3 | NP_110446.3 | 2 | 467 | |||
XM_017023738.1 | XP_016879227.1 | 2 | 417 | |||
GenBank mRNA | AK074382.1 | - | 3 | 420 | ||
AK225997.1 | - | 2 | 437 | |||
AK307280.1 | - | 2 | 421 | |||
AY358169.1 | - | 1 | 177 | |||
BC002943.1 | - | 1 | 94 | |||
BC107585.1 | - | 2 | 417 | 6463662 | ||
BI826603.1 | - | 2 | 518 | |||
DC368734.1 | - | 2 | 467 | |||
DC381447.1 | - | 2 | 422 |