Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032134.2 | NP_115510.1 | 2, 3 | 275 | ||
NR_130649.1 | 2, 3 | 275 | ||||
XM_005257728.3 | XP_005257785.1 | 2, 3 | 1285 | |||
XM_017025206.1 | XP_016880695.1 | 2, 3 | 1285 | |||
XM_017025208.1 | XP_016880697.1 | 1, 2 | 363 | |||
GenBank mRNA | AL136774.1 | - | 2, 3 | 275 | ||
BC131559.1 | - | 2, 3 | 160 |